accessibility menu, dialog, popup

UserName

CAS RN 63020-83-7

HNNOOH(E/Z)

CAS RN 63020-83-7

IUPAC Name: 4-[2-(9H-fluoren-2-yl)hydrazin-1-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
Synonyms: 4-[2-(9H-fluoren-2-yl)hydrazin-1-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 302.33
Molecular Formula: C19H14N2O2
InChi Key: SNNPROHVBXRSHJ-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C=CC1=NNC1=CC2=C(C=C1)C1=CC=CC=C1C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results