missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-2-azo-2',4'-dihydroxybenzene 98.0+%, TCI America™

Catalog No. F00191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F00191G Supplier TCI America Supplier No. F00191G
Only null left

Chemical Identifiers

CAS 63020-83-7
Molecular Formula C19H14N2O2
Molecular Weight (g/mol) 302.333
MDL Number MFCD00059493
InChI Key SNNPROHVBXRSHJ-UHFFFAOYSA-N
Synonym 4-(2-Fluorenylazo)resorcinol
PubChem CID 54202147
IUPAC Name 4-(9H-fluoren-2-ylhydrazinylidene)-3-hydroxycyclohexa-2,5-dien-1-one
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)NN=C4C=CC(=O)C=C4O

Specifications

Color Red
Quantity 1 g
Formula Weight 302.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Fluorene-2-azo-2′,4′-dihydroxybenzene