accessibility menu, dialog, popup

UserName

H3COH

CAS RN 629-76-5

IUPAC Name: pentadecan-1-ol
Synonyms: 1-pentadecanol
Molecular Weight (g/mol): 228.42
Molecular Formula: C15H32O
InChi Key: REIUXOLGHVXAEO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCO
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)