missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecanol, 99%

Catalog No. AAL0367306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0367306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0367306 Supplier Thermo Scientific Chemicals Supplier No. L0367306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-76-5 | C15H32O | 228.42 g/mol

1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing a

Chemical Identifiers

CAS 629-76-5
Molecular Formula C15H32O
Molecular Weight (g/mol) 228.42
MDL Number MFCD00004759
InChI Key REIUXOLGHVXAEO-UHFFFAOYSA-N
Synonym 1-pentadecanol, pentadecyl alcohol, pentadecanol, n-pentadecanol, n-1-pentadecanol, neodol 5, unii-333qva4g2q, dsstox_cid_7270, dsstox_rid_78381, dsstox_gsid_27270
PubChem CID 12397
ChEBI CHEBI:77468
IUPAC Name pentadecan-1-ol
SMILES CCCCCCCCCCCCCCCO

Specifications

Melting Point 45°C to 46°C
Boiling Point 269°C to 271°C
Quantity 5 g
Beilstein 1700695
Solubility Information Insoluble in water.
Formula Weight 228.42
Percent Purity 99%
Chemical Name or Material 1-Pentadecanol
EINECSNumber 211-107-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only