missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecanol 95.0+%, TCI America™

Catalog No. P0036500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0036500G 500 g
P003625G 25 g
P0036100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0036500G Supplier TCI America Supplier No. P0036500G

Chemical Identifiers

CAS 629-76-5
Molecular Formula C15H32O
Molecular Weight (g/mol) 228.42
MDL Number MFCD00004759
InChI Key REIUXOLGHVXAEO-UHFFFAOYSA-N
Synonym 1-pentadecanol, pentadecyl alcohol, pentadecanol, n-pentadecanol, n-1-pentadecanol, neodol 5, unii-333qva4g2q, dsstox_cid_7270, dsstox_rid_78381, dsstox_gsid_27270
PubChem CID 12397
ChEBI CHEBI:77468
IUPAC Name pentadecan-1-ol
SMILES CCCCCCCCCCCCCCCO

Specifications

Melting Point 44°C
Color White
Boiling Point 133°C
Quantity 500 g
Formula Weight 228.42
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Pentadecanol
EINECSNumber (2)-0217
TSCA Yes