accessibility menu, dialog, popup

UserName

CAS RN 6287-90-7

H3COOBr

CAS RN 6287-90-7

IUPAC Name: methyl 11-bromoundecanoate
Synonyms: methyl 11-bromoundecanoate
Molecular Weight (g/mol): 279.22
Molecular Formula: C12H23BrO2
InChi Key: HFNPVFKUZYCDIB-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results