accessibility menu, dialog, popup

UserName

ONHHNO

CAS RN 628-36-4

IUPAC Name: N'-formylformohydrazide
Synonyms: diformylhydrazine
Molecular Weight (g/mol): 88.07
Molecular Formula: C2H4N2O2
InChi Key: POVXOWVFLAAVBH-UHFFFAOYSA-N
SMILES: O=CNNC=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)