missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diformylhydrazine, TCI America™

Catalog No. D459525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D459525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D459525G Supplier TCI America Supplier No. D459525G

Chemical Identifiers

CAS 628-36-4
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.07
MDL Number MFCD00003275
InChI Key POVXOWVFLAAVBH-UHFFFAOYSA-N
Synonym 1,2-diformylhydrazine, n'-formylformohydrazide, diformylhydrazine, 1,2-hydrazinedicarboxaldehyde, bicarbamaldehyde, diformohydrazide, s-diformohydrazide, s-diformylhydrazine, n,n'-diformylhydrazine, n'-formylformic hydrazide
PubChem CID 12342
IUPAC Name N'-formylformohydrazide
SMILES O=CNNC=O

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 88.07
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Diformylhydrazine
RTECSNumber LQ8625000
TSCA Yes