missing translation for 'onlineSavingsMsg'
Learn More

sym-Diformylhydrazine, 97%, Thermo Scientific Chemicals

Catalog No. AC114640250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114640250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC114640250 Supplier Thermo Scientific Chemicals Supplier No. 114640250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-36-4 | Molecular Formula: C2H4N2O2 | Molecular Weight: 88.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-36-4
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.07
MDL Number MFCD00003275
InChI Key POVXOWVFLAAVBH-UHFFFAOYSA-N
Synonym 1,2-diformylhydrazine, n'-formylformohydrazide, diformylhydrazine, 1,2-hydrazinedicarboxaldehyde, bicarbamaldehyde, diformohydrazide, s-diformohydrazide, s-diformylhydrazine, n,n'-diformylhydrazine, n'-formylformic hydrazide
PubChem CID 12342
SMILES O=CNNC=O

Specifications

Melting Point 155°C to 160°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HCONHNHCHO
Quantity 25 g
Beilstein 02, 93
IUPAC Name N-formamidoformamide
Formula Weight 88.06
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material sym-Diformylhydrazine, 97%
Show More Show Less
EINECSNumber 211-040-5
RTECSNumber LQ8625000
TSCA TSCA

RUO – Research Use Only