accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 628-02-4

IUPAC Name: hexanamide
Synonyms: nylon 6 nylon-6
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ALBYIUDWACNRRB-UHFFFAOYSA-N
SMILES: CCCCCC(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)