missing translation for 'onlineSavingsMsg'
Learn More

Hexanamide 98.0+%, TCI America™

Catalog No. C00245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00245G 5 g
C002425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00245G Supplier TCI America Supplier No. C00245G

Chemical Identifiers

CAS 628-02-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00008046
InChI Key ALBYIUDWACNRRB-UHFFFAOYSA-N
Synonym capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide
PubChem CID 12332
IUPAC Name hexanamide
SMILES CCCCCC(=O)N

Specifications

Melting Point 100°C
Color White
Quantity 5 g
Formula Weight 115.18
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexanamide
EINECSNumber (2)-0824
RTECSNumber MN7875000
TSCA Yes