missing translation for 'onlineSavingsMsg'
Learn More

Hexanamide, 98%, Thermo Scientific™

Catalog No. AC411530010
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411530010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC411530010 Supplier Thermo Scientific Chemicals Supplier No. 411530010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-02-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008046
InChI Key ALBYIUDWACNRRB-UHFFFAOYSA-N
Synonym capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide
PubChem CID 12332
IUPAC Name hexanamide
SMILES CCCCCC(=O)N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4CONH2
Quantity 1g
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material Hexanamide, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-024-8
RTECSNumber MN7875000
TSCA TSCA

RUO – Research Use Only