accessibility menu, dialog, popup

UserName

CAS RN 627-01-0

H3CNH2OO

CAS RN 627-01-0

IUPAC Name: 2-(ethylazaniumyl)acetate
Synonyms: (ethylammonio)acetate N-ethylglycine zwitterion
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: YPIGGYHFMKJNKV-UHFFFAOYSA-N
SMILES: CC[NH2+]CC([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results