missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylglycine 98.0+%, TCI America™

$180.97 - $1678.75

Chemical Identifiers

CAS 627-01-0
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00037794
InChI Key YPIGGYHFMKJNKV-UHFFFAOYSA-N
Synonym n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa
PubChem CID 316542
ChEBI CHEBI:15620
IUPAC Name 2-(ethylamino)acetic acid
SMILES CCNCC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E05171G
View Documents
TCI America
E05171G
1 g
Each for $180.97
Only null left
 
E05175G
View Documents
TCI America
E05175G
5 g
Each for $487.07
Only null left
 
E051725G
View Documents
TCI America
E051725G
25 g
Each for $1,678.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

627-01-0
103.121
YPIGGYHFMKJNKV-UHFFFAOYSA-N
316542
2-(ethylamino)acetic acid
C4H9NO2
MFCD00037794
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa
CHEBI:15620
CCNCC(=O)O

Specifications

627-01-0
White-Yellow
MFCD00037794
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa
CCNCC(=O)O
103.121
CHEBI:15620
≥98.0% (T)
N-Ethylglycine
190°C
C4H9NO2
1 g
YPIGGYHFMKJNKV-UHFFFAOYSA-N
2-(ethylamino)acetic acid
316542
103.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'