accessibility menu, dialog, popup

UserName

CAS RN 62625-16-5

IUPAC Name: tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Synonyms: tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Molecular Weight (g/mol): 610.73
Molecular Formula: C20H13K3O11S2
InChi Key: PLHSVCWOEHYSHO-UHFFFAOYSA-K
SMILES: [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)