missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein disulfate tripotassium salt hydrate, 98%

Catalog No. AAB2419806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2419806 5 g
AAB2419803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2419806 Supplier Thermo Scientific Chemicals Supplier No. B2419806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-16-5 | Molecular Formula: C20H13K3O11S2 | Molecular Weight: 610.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-16-5
Molecular Formula C20H13K3O11S2
Molecular Weight (g/mol) 610.73
MDL Number MFCD00150194
InChI Key PLHSVCWOEHYSHO-UHFFFAOYSA-K
Synonym phenolphthalein disulfate potassium salt, phenolphthalein disulfate tripotassium salt hydrate
PubChem CID 129893851
SMILES [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O

Specifications

Quantity 5 g
IUPAC Name tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Formula Weight 610.75 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Phenolphthalein disulfate tripotassium salt hydrate
EINECSNumber 263-648-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only