missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 230.0°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Assay Percent Range | 98% |
| Color | White |
| Solubility Information | Solubility in water: slightly soluble. |
| Quantity | 10 g |
| Physical Form | Crystalline Powder |
| IUPAC Name | 2-[hydroxy-bis(4-sulfooxyphenyl)methyl]benzoic acid;potassium |
| Formula Weight | 664.78 |
| Show More |
Safety and Handling
| EINECSNumber | 263-648-5 |
| TSCA | TSCA |
RUO – Research Use Only