accessibility menu, dialog, popup

UserName

CH3Cl(E/Z)

CAS RN 62614-70-4

IUPAC Name: 6-chlorohex-2-ene
Synonyms: 6-chlorohex-2-ene
Molecular Weight (g/mol): 118.6
Molecular Formula: C6H11Cl
InChi Key: SJHDJTNQXRYLLN-UHFFFAOYSA-N
SMILES: CC=CCCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results