missing translation for 'onlineSavingsMsg'
Learn More

cis-6-Chloro-2-hexene 90.0+%, TCI America™

Catalog No. C16535ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16535ML 5 mL
C165325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16535ML Supplier TCI America Supplier No. C16535ML
Only null left

Chemical Identifiers

CAS 62614-70-4
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD02093458
InChI Key SJHDJTNQXRYLLN-IHWYPQMZSA-N
Synonym cis-4-Hexen-1-yl Chloride
PubChem CID 12466001
IUPAC Name (Z)-6-chlorohex-2-ene
SMILES CC=CCCCCl

Specifications

Color Colorless
Boiling Point 132°C
Quantity 5 mL
UN Number 1993
Formula Weight 118.60
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-6-Chloro-2-hexene