accessibility menu, dialog, popup

UserName

CH3(S)HN

CAS RN 626-56-2

IUPAC Name: (3S)-3-methylpiperidine
Synonyms: (3S)-3-methylpiperidine
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: JEGMWWXJUXDNJN-LURJTMIESA-N
SMILES: C[C@H]1CCCNC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)