missing translation for 'onlineSavingsMsg'
Learn More

3-Méthylpipéridine 97,0+%, TCI America™

Numéro de catalogue. P0444500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P0444500ML 500 mL
P044425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P0444500ML Fournisseur TCI America Code fournisseur P0444500ML

Identifiants chimiques

CAS 626-56-2
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005994
InChI Key JEGMWWXJUXDNJN-UHFFFAOYSA-N
Synonym 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc
PubChem CID 79081
IUPAC Name 3-méthylpipéridine
SMILES CC1CCCNC1

Spécifications

Color Jaune
Boiling Point 126°C
Quantity 500 mL
UN Number 1993
Formula Weight 99.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquide
Chemical Name or Material 3-Methylpiperidine
EINECSNumber (5)-0768
TSCA Yes