missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpiperidine, 97%

Catalog No. AAL1340822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1340822 100 g
AAL1340814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1340822 Supplier Thermo Scientific Chemicals Supplier No. L1340822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 626-56-2 | C6H13N | 99.177 g/mol

3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.

Chemical Identifiers

CAS 626-56-2
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005994
InChI Key JEGMWWXJUXDNJN-UHFFFAOYSA-N
Synonym 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc
PubChem CID 79081
IUPAC Name 3-methylpiperidine
SMILES CC1CCCNC1

Specifications

Melting Point -24°C
Density 0.845
Boiling Point 124°C to 125°C
Flash Point 8°C (46°F)
Odor Amine-like
Refractive Index 1.447
Quantity 100 g
UN Number UN2733
Beilstein 79807
Solubility Information It is soluble in water.
Formula Weight 99.18
Percent Purity 97%
Chemical Name or Material 3-Methylpiperidine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-953-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only