accessibility menu, dialog, popup

UserName

CAS RN 626-55-1

IUPAC Name: 3-bromopyridine
Synonyms: 3-bromopyridine
Molecular Weight (g/mol): 158
Molecular Formula: C5H4BrN
InChi Key: NYPYPOZNGOXYSU-UHFFFAOYSA-N
SMILES: BrC1=CC=CN=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)