missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine, 98+%

Catalog No. AAA1289436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1289436 500 g
AAA1289414 25 g
AAA1289422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1289436 Supplier Thermo Scientific Chemicals Supplier No. A1289436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-55-1 | C5H4BrN | 158.00 g/mol

3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.

Solubility
Soluble in water (31 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.

Chemical Identifiers

CAS 626-55-1
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 158.00
MDL Number MFCD00006373
InChI Key NYPYPOZNGOXYSU-UHFFFAOYSA-N
Synonym 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
PubChem CID 12286
ChEBI CHEBI:51575
IUPAC Name 3-bromopyridine
SMILES BrC1=CC=CN=C1

Specifications

Melting Point -27°C
Density 1.617
Boiling Point 172°C to 174°C
Flash Point 72°C (181°F)
Odor Characteristic
Refractive Index 1.57
Quantity 500 g
UN Number UN2810
Beilstein 105880
Solubility Information Soluble in water (31g/L at 20°C).
Formula Weight 158
Percent Purity ≥98%
Chemical Name or Material 3-Bromopyridine
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-952-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only