missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. B065125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B065125G 25 g
B0651100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B065125G Supplier TCI America Supplier No. B065125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-55-1
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 158.00
MDL Number MFCD00006373
InChI Key NYPYPOZNGOXYSU-UHFFFAOYSA-N
Synonym 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
PubChem CID 12286
ChEBI CHEBI:51575
IUPAC Name 3-bromopyridine
SMILES BrC1=CC=CN=C1

Specifications

Melting Point -27°C
Color Yellow
Boiling Point 178°C
Quantity 25 g
UN Number 1993
Formula Weight 158.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromopyridine (stabilized with Copper chip)