CAS RN 6259-42-3
IUPAC Name:
hydrogen 5-chloro-2-methylaniline chloride
Synonyms:
hydrogen 5-chloro-O-toluidine chloride
Molecular Weight (g/mol):
178.06
Molecular Formula:
C7H9Cl2N
InChi Key:
BMZGSMUCRXYUGB-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=CC=C(Cl)C=C1N
1
–
1
of
1
results
1
5-Chloro-2-methylaniline Hydrochloride 98.0+%, TCI America™
CAS: 6259-42-3 Molecular Formula: C7H9Cl2N Molecular Weight (g/mol): 178.056 MDL Number: MFCD00035487 InChI Key: BMZGSMUCRXYUGB-UHFFFAOYSA-N Synonym: 2-Amino-4-chlorotoluene Hydrochloride, 5-Chloro-o-toluidine Hydrochloride PubChem CID: 80416 IUPAC Name: 5-chloro-2-methylaniline;hydrochloride SMILES: CC1=C(C=C(C=C1)Cl)N.Cl