missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine 98.0+%, TCI America™

Catalog No. D25735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25735G 5 g
D257325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25735G Supplier TCI America Supplier No. D25735G

Chemical Identifiers

CAS 625-92-3
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00014634
InChI Key SOSPMXMEOFGPIM-UHFFFAOYSA-N
Synonym pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
PubChem CID 69369
ChEBI CHEBI:51593
IUPAC Name 3,5-dibromopyridine
SMILES BrC1=CC(Br)=CN=C1

Specifications

Melting Point 111°C
Color White-Yellow
Boiling Point 222°C
Quantity 5 g
Formula Weight 236.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromopyridine