missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine, 98+%

Catalog No. AC330131000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330131000 100 g Glass bottle
AC330130100 10 g Glass bottle
AC330130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC330131000 Supplier Thermo Scientific Chemicals Supplier No. 330131000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 625-92-3 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-92-3
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00014634
InChI Key SOSPMXMEOFGPIM-UHFFFAOYSA-N
Synonym pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
PubChem CID 69369
ChEBI CHEBI:51593
IUPAC Name 3,5-dibromopyridine
SMILES BrC1=CC(Br)=CN=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 110°C to 114°C
Color White to Beige
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 236.91
Percent Purity 98+%
Physical Form Needle-like Crystals
Chemical Name or Material 3, 5-Dibromopyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-916-4

RUO – Research Use Only