accessibility menu, dialog, popup

UserName

CH3H2NCH3H3C

CAS RN 625-43-4

IUPAC Name: methyl(2-methylpropyl)azanium
Synonyms: methyl(2-methylpropyl)azanium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: QKYWADPCTHTJHQ-UHFFFAOYSA-O
SMILES: C[NH2+]CC(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)