missing translation for 'onlineSavingsMsg'
Learn More

N-Methylisobutylamine, 98+%

Catalog No. AA4294706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4294706 5 g
AA4294703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4294706 Supplier Thermo Scientific Chemicals Supplier No. 04294706

CAS: 625-43-4 | C5H13N | 87.166 g/mol

N-Methylisobutylamine is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-43-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015043
InChI Key QKYWADPCTHTJHQ-UHFFFAOYSA-N
Synonym n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin
PubChem CID 12249
IUPAC Name N,2-dimethylpropan-1-amine
SMILES CC(C)CNC

Specifications

Density 0.73 g/mL
Boiling Point 78°C
Odor Amine-like
Quantity 5 g
UN Number UN2733
Solubility Information Miscible with water.
Formula Weight 87.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N-Methylisobutylamine
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-893-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only