accessibility menu, dialog, popup

UserName

CH3NHCH3

CAS RN 622-57-1

IUPAC Name: N-ethyl-4-methylaniline
Synonyms: benzenamine, N-ethyl-4-methyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: AASABFUMCBTXRL-UHFFFAOYSA-N
SMILES: CCNC1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)