accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 62-75-9

CH3NH3CNO

CAS RN 62-75-9

IUPAC Name: dimethyl(nitroso)amine
Synonyms: nitrosodimethylamine DMNA ndma nitrosodimethylamine rcra waste number P082 DMN nitrosodimethylamine, ndma ccris 261 N-nitroaodimethylamine
Molecular Weight (g/mol): 74.08
Molecular Formula: C2H6N2O
InChi Key: UMFJAHHVKNCGLG-UHFFFAOYSA-N
SMILES: CN(C)N=O
Grade 
  • (1)
  • (1)
Concentration 
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.