missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ N-Nitrosodimethylamine, MilliporeSigma™ Supelco™

Catalog No. 111017891
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017891 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017891 Supplier Merck Emd Millipore Supplier No. CRM40059
Only null left

Specifications

Packaging Ampule
CAS 62-75-9
Product Line TraceCERT
Concentration 5000 μg/mL in methanol
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC