accessibility menu, dialog, popup

UserName

CAS RN 62-32-8

IUPAC Name: hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
Synonyms: hydrogen methyldopamine chloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: JCDRZCWRRLKLTB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results