accessibility menu, dialog, popup

UserName

CAS RN 619-55-6

IUPAC Name: 4-methylbenzamide
Synonyms: 4-methylbenzamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: UHBGYFCCKRAEHA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(N)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)