accessibility menu, dialog, popup

UserName

CAS RN 6184-17-4

NNNHNP(S)(S)(R)OOOOOOHOHOHOHCH3CH3NaH2OH2O

CAS RN 6184-17-4

IUPAC Name: sodium 10-[(2S,3S,4R)-5-(hydrogen phosphonatooxy)-2,3,4-trihydroxypentyl]-7,8-dimethyl-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione dihydrate
Synonyms: sodium 10-[(2S,3S,4R)-5-(hydrogen phosphonatooxy)-2,3,4-trihydroxypentyl]-7,8-dimethyl-3H-benzo[g]pteridine-2,4-dione dihydrate
Molecular Weight (g/mol): 514.36
Molecular Formula: C17H24N4NaO11P
InChi Key: LQVGUKOCMOKKJU-FTNKLFHDNA-M
SMILES: O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results