CAS RN 618-98-4
IUPAC Name:
ethyl 3-nitrobenzoate
Synonyms:
ethyl-M-nitrobenzoate
Molecular Weight (g/mol):
195.17
Molecular Formula:
C9H9NO4
InChi Key:
MKBIJCPQTPFQKQ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-nitrobenzoate, 98+%
Ethyl 3-nitrobenzoate, 98+%
CAS: 618-98-4 Molecular Formula: C9H9NO4 Molecular Weight (g/mol): 195.174 MDL Number: MFCD00014702 InChI Key: MKBIJCPQTPFQKQ-UHFFFAOYSA-N Synonym: ethyl m-nitrobenzoate,benzoic acid, 3-nitro-, ethyl ester,ethyl-m-nitrobenzoate,m-nitrobenzoic acid, ethyl ester,benzoic acid, m-nitro-, ethyl ester,acmc-20akzc,3-nitrobenzoic acid ethyl,benzoic acid, 3-nitro-,ethyl ester,benzoic acid, m-nitro-, ethyl ester 8ci,r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline PubChem CID: 69261 IUPAC Name: ethyl 3-nitrobenzoate SMILES: CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-]