accessibility menu, dialog, popup

UserName

CAS RN 618-62-2

IUPAC Name: 1,3-dichloro-5-nitrobenzene
Synonyms: benzene, 1,3-dichloro-5-nitro-
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: RNABGKOKSBUFHW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results