accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 616-75-1

IUPAC Name: 2-benzylpropanedioic acid
Synonyms: 2-benzylmalonate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: JAEJSNFTJMYIEF-UHFFFAOYSA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results