accessibility menu, dialog, popup

UserName

CAS RN 616-74-0

IUPAC Name: 4,6-dinitrobenzene-1,3-diol
Synonyms: 1,3-benzenediol, 4,6-dinitro-
Molecular Weight (g/mol): 200.11
Molecular Formula: C6H4N2O6
InChi Key: MBXIRNHACKAGPA-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results