accessibility menu, dialog, popup

UserName

CH3NNO

CAS RN 614-00-6

IUPAC Name: N-methyl-N-nitrosoaniline
Synonyms: N-methyl-N-nitrosobenzeneamine
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: MAXCWSIJKVASQC-UHFFFAOYSA-N
SMILES: CN(N=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results