accessibility menu, dialog, popup

UserName

CAS RN 61367-07-5

IUPAC Name: methyl 4-aminocyclohexane-1-carboxylate hydrochloride
Synonyms: methyl 4-aminocyclohexane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C8H16ClNO2
InChi Key: NHAYDXCUCXRAMF-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1CCC(N)CC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)