missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride, 97%

Catalog No. AAH6256206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6256206 5 g
AAH6256203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6256206 Supplier Thermo Scientific Chemicals Supplier No. H6256206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61367-07-5 | C8H16ClNO2 | 193.671 g/mol

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 61367-07-5
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
MDL Number MFCD08274538
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym methyl trans-4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl cis-4-aminocyclohexanecarboxylate hydrochloride, cis-methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexane-1-carboxylate hydrochloride, trans-4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride, cyclohexanecarboxylic acid, 4-amino-, methyl ester, hydrochloride, trans-4-amino-cyclohexylcarboxylic acid methyl ester hydrochloride, trans-methyl 4-aminocyclohexanecarboxylate hydrochloride, 4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl

Specifications

Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 193.67
Percent Purity 97%
Chemical Name or Material Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only