accessibility menu, dialog, popup

UserName

CAS RN 6117-91-5

CH3OH(Z)

CAS RN 6117-91-5

IUPAC Name: (2Z)-but-2-en-1-ol
Synonyms: crotyl alcohol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: WCASXYBKJHWFMY-IHWYPQMZSA-N
SMILES: C\C=C/CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results