missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotyl Alcohol (cis- and trans- mixture) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 121°C |
| Quantity | 25 mL |
| UN Number | 1987 |
| Formula Weight | 72.11 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Crotyl Alcohol (cis- and trans- mixture) |
Safety and Handling
| RTECSNumber | EM9275000 |
| TSCA | Yes |