accessibility menu, dialog, popup

UserName

HOOO

CAS RN 611-95-0

IUPAC Name: 4-benzoylbenzoic acid
Synonyms: benzoic acid, 4-benzoyl-
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: IFQUPKAISSPFTE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)