missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbenzoic acid, 99%, Thermo Scientific™

Catalog No. p-3777393
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105720050 5g Glass bottle
AC105720250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105720050 Supplier Thermo Scientific Chemicals Supplier No. 105720050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 611-95-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002560
InChI Key IFQUPKAISSPFTE-UHFFFAOYSA-N
Synonym p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
PubChem CID 69147
IUPAC Name 4-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5COC6H4CO2H
Quantity 5g
Beilstein 10, 753
Formula Weight 226.23
Percent Purity 99%
Chemical Name or Material 4-Benzoylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-286-

RUO – Research Use Only