CAS RN 610769-35-2
IUPAC Name:
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol; boric acid
Synonyms:
boric acid; edetic acid; tris buffer
Molecular Weight (g/mol):
475.21
Molecular Formula:
C14H30BN3O14
InChi Key:
OSBLTNPMIGYQGY-UHFFFAOYSA-N
SMILES:
OB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Thermo Scientific Chemicals Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
CAS: 610769-35-2 Molecular Formula: C14H30BN3O14 Molecular Weight (g/mol): 475.211 MDL Number: MFCD00236358 InChI Key: OSBLTNPMIGYQGY-UHFFFAOYSA-N Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol,TRIS,Tris buffer,Tromethamine,Tromethane PubChem CID: 21257725 IUPAC Name: 2-amino-2-(hydroxymethyl)propane-1,3-diol;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;boric acid SMILES: B(O)(O)O.C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.C(C(CO)(CO)N)O