accessibility menu, dialog, popup

UserName

CAS RN 610-96-8

IUPAC Name: methyl 2-chlorobenzoate
Synonyms: methyl O-chlorobenzoate
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: JAVRNIFMYIJXIE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1Cl
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)