accessibility menu, dialog, popup

UserName

CAS RN 609-93-8

IUPAC Name: 4-methyl-2,6-dinitrophenol
Synonyms: victoria yellow
Molecular Weight (g/mol): 198.13
Molecular Formula: C7H6N2O5
InChi Key: HOYRZHJJAHRMLL-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results